Assay Details
| Gene Symbol: 2010012O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025563.3 | NP_079839.1 | 1 - 2 | 179 | 80 | |
| GenBank mRNA | AK007626.1 | - | 1 - 2 | 166 | 80 | |
| AK008215.1 | - | 1 - 2 | 174 | 80 | ||
| AK046524.1 | - | 1 - 2 | 179 | 80 | ||
| BC027506.1 | - | 1 - 2 | 151 | 1514880 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145406 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145405 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145407 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145408 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | AK046524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | BC027506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | NM_025563.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | AK008215.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | AK046524.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | BC027506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | NM_025563.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267508 | AK008215.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130305 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172769 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174214 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | AK046524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | BC027506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | NM_025563.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | AK008215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | AK046524.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | BC027506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | NM_025563.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267508 | AK008215.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32145384 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174214 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03607332 | - | - |
| Whole Mouse Genome G4122A | A_52_P386302 | AK046524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P386302 | BC027506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P386302 | NM_025563.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P386302 | AK007626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P386302 | AK008215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P635110 | AK046524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P635110 | NM_025563.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | AK046524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | BC027506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | NM_025563.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | AK008215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | AK046524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | BC027506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | NM_025563.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267508 | AK008215.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135235 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120754 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174214 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14389607 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14337981 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14337983 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14381981 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14389604 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14389605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145406 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145407 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145405 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32145409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32145369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14462422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14187258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14481771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14326312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14389601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14173697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14389610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14173694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14183202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14337983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14389605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32145431 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10463702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10463697 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419299_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439561_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10463695 | - | - |
| Mouse Genome 430 2.0 Array | 1439561_at | - | - |
| Mouse Genome 430 2.0 Array | 1419299_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419299_at | - | - |
| Mouse Genome 430 2.0 Array | 1439561_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66439_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66439_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900006300.mm.1 | NM_025563.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006300.mm.1 | NM_025563.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006300.mm.1 | TC1900000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900006302.mm.1 | NM_025563.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006302.mm.1 | NM_025563.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006302.mm.1 | TC1900000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900006304.mm.1 | NM_025563.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006304.mm.1 | NM_025563.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900006304.mm.1 | TC1900000708.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419299_at | - | - |
| Mouse Expression Set 430 | 1419299_PM_at | - | - |
| Mouse Expression Set 430 | 1439561_PM_at | - | - |
| Mouse Expression Set 430 | 1439561_at | - | - |
| Murine Genome U74v2 Set | 168094_PM_i_at | - | - |
| Murine Genome U74v2 Set | 128689_at | - | - |
| Murine Genome U74v2 Set | 168094_i_at | - | - |
| Murine Genome U74v2 Set | 128689_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419299_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419299_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419299_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419299_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439561_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439561_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 831483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480086 | - | - |
| MouseWG-6 v2 Expression BeadChip | 103850167 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103850167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2748300 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2748300 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246171 | - | - |
| mouse-6 v1.1 (Illumina) | scl6004.1.1_83-S | - | - |
| mouse-6 v1.1 (Illumina) | scl53052.6_95-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103850167 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53052.6_95-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480086 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103850167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2748300 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2748300 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53052.6_95-S | - | - |

2010012O05Rik
Genomic Map