Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081188.1 | NP_001074657.1 | 4 - 5 | 456 | 65 | |
| GenBank mRNA | AK011294.1 | - | 4 - 5 | 443 | 65 | |
| AK129057.1 | - | 4 - 5 | 408 | 65 | ||
| BC039655.1 | - | 4 - 5 | 441 | 65 | ||
| BC052656.1 | - | 4 - 5 | 438 | 30041585 | 65 | |
| BC094932.1 | - | 4 - 5 | 449 | 30008071 | 65 | |
| BC132363.1 | - | 4 - 5 | 471 | 40130640 | 65 | |
| BC145675.1 | - | 4 - 5 | 471 | 40130978 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074771 | BC052656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074771 | BC094932.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074771 | BC039655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074771 | AK129057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074771 | NM_001081188.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 1318 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134871 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406788 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 6317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68636 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146033 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154459 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106456 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074771 | BC052656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074771 | BC094932.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074771 | BC039655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074771 | AK129057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074771 | NM_001081188.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146033 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154459 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02050895 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106456 | - | - |
| Whole Mouse Genome G4122A | A_51_P381784 | BC094932.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381784 | BC132363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381784 | BC039655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381784 | BC145675.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381784 | AK129057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P296048 | AK085426.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06956067 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106456 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154459 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146033 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07917018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07493368 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07917009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07917016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26893323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893231 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08014547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07882812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07917018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07988338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07934215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07882808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08168961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07882807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07732722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26893357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08014543 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10590551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590555 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590552 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590553 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10590556 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448365_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10590549 | - | - |
| Mouse Genome 430 2.0 Array | 1448365_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448365_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66446_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66446_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900013938.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013938.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013938.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013930.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013930.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013930.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013936.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013936.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013936.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013929.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013929.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013929.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013939.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013939.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013939.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013926.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013926.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013926.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013931.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013931.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013931.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013935.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013935.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013935.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013934.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013934.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013934.mm.1 | TC0900001656.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900013940.mm.1 | NM_001081188.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013940.mm.1 | NM_001081188.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900013940.mm.1 | TC0900001656.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448365_PM_at | - | - |
| Mouse Expression Set 430 | 1448365_at | - | - |
| Murine Genome U74v2 Set | 99151_at | - | - |
| Murine Genome U74v2 Set | 99151_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448365_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448365_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448365_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448365_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 453158 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105340040 | - | - |
| MouseWG-6 v2 Expression BeadChip | 520538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105340040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2715289 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223393 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2715289 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105340040 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105340040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2715289 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223393 | - | - |

Exosc7
Genomic Map