Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081188.1 | NP_001074657.1 | 5 - 6 | 524 | 95 | |
| GenBank mRNA | AK004437.1 | - | 2 - 3 | 96 | 95 | |
| AK011294.1 | - | 5 - 6 | 511 | 95 | ||
| AK129057.1 | - | 5 - 6 | 476 | 95 | ||
| BC039655.1 | - | 5 - 6 | 509 | 95 | ||
| BC094932.1 | - | 5 - 6 | 517 | 30008071 | 95 | |
| BC132363.1 | - | 5 - 6 | 539 | 40130640 | 95 | |
| BC145675.1 | - | 5 - 6 | 539 | 40130978 | 95 | |
More Information
Set Membership: |
|

Exosc7
Genomic Map