Assay Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025626.4 | NP_079902.1 | 2 - 3 | 365 | 73 | |
| GenBank mRNA | AK013932.2 | - | 2 - 3 | 330 | 73 | |
| AK019815.1 | - | 2 - 3 | 277 | 73 | ||
| AK032875.1 | - | 2 - 3 | 366 | 73 | ||
| AK168759.1 | - | 2 - 3 | 331 | 73 | ||
| BC021353.1 | - | 2 - 3 | 331 | 5006672 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam107b
Genomic Map