Assay Details
| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025962.3 | NP_080238.2 | 2 - 3 | 348 | 92 | |
| GenBank mRNA | AK007725.1 | - | 2 - 3 | 283 | 92 | |
| AK012761.1 | - | 2 - 3 | 285 | 92 | ||
| AK084194.1 | - | 2 - 3 | 299 | 92 | ||
| AK213975.1 | - | 1 - 2 | 49 | 92 | ||
| BB614283.1 | - | 2 - 3 | 349 | 92 | ||
| BC054756.1 | - | 2 - 3 | 287 | 6411797 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmachc
Genomic Map