Assay Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025982.4 | NP_080258.1 | 3 - 4 | 400 | 70 | |
| GenBank mRNA | AK012457.1 | - | 3 - 4 | 401 | 70 | |
| AK012567.1 | - | 3 - 4 | 403 | 70 | ||
| AK012853.1 | - | 3 - 4 | 403 | 70 | ||
| AK013523.1 | - | 3 - 4 | 413 | 70 | ||
| AK014183.1 | - | 3 - 4 | 399 | 70 | ||
| AK075962.1 | - | 3 - 4 | 401 | 70 | ||
| AK178759.1 | - | 2 - 3 | 138 | 70 | ||
| AK180941.1 | - | 2 - 3 | 138 | 70 | ||
| AK183488.1 | - | 2 - 3 | 116 | 70 | ||
| BC023306.1 | - | 3 - 4 | 377 | 4165774 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan31
Genomic Map