Assay Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025982.4 | NP_080258.1 | 5 - 6 | 649 | 130 | |
| GenBank mRNA | AK012457.1 | - | 5 - 6 | 650 | 130 | |
| AK012567.1 | - | 5 - 6 | 652 | 130 | ||
| AK012853.1 | - | 5 - 6 | 652 | 130 | ||
| AK014183.1 | - | 5 - 6 | 648 | 130 | ||
| AK075962.1 | - | 5 - 6 | 650 | 130 | ||
| BC023306.1 | - | 5 - 6 | 626 | 4165774 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407074 | AK012457.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P407074 | AK012457.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814468 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134538 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407074 | AK012457.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P407074 | AK012457.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27533859 | - | - |
| Whole Mouse Genome G4122A | A_51_P407074 | AK012457.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P407074 | AK012457.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133053 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08437198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08437200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08437201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08323692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27533843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08437200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08750467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08903907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08606902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08323680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08323692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08501998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08380748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27533879 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373028 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373035 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373034 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455853_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430029_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416556_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373027 | - | - |
| Mouse Genome 430 2.0 Array | 1416556_at | - | - |
| Mouse Genome 430 2.0 Array | 1455853_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430029_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430029_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455853_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1416556_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67125_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67125_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023044.mm.1 | NM_025982.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023044.mm.1 | NM_025982.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023044.mm.1 | TC1000003096.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430029_a_at | - | - |
| Mouse Expression Set 430 | 1430029_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416556_at | - | - |
| Mouse Expression Set 430 | 1455853_x_at | - | - |
| Mouse Expression Set 430 | 1416556_PM_at | - | - |
| Mouse Expression Set 430 | 1455853_PM_x_at | - | - |
| Murine Genome U74v2 Set | 98144_f_at | - | - |
| Murine Genome U74v2 Set | 98144_PM_f_at | - | - |
| Murine Genome U74v2 Set | 170314_PM_f_at | - | - |
| Murine Genome U74v2 Set | 170314_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430029_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455853_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455853_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416556_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416556_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430029_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430029_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430029_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455853_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455853_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416556_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416556_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2030333 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520279 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2030333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520279 | - | - |
| mouse-6 v1.1 (Illumina) | scl37402.3_73-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030333 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520279 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067125.2_158-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37402.3_73-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030333 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520279 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2964076 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2757599 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2964076 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757599 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37402.3_73-S | - | - |

Tspan31
Genomic Map