Assay Details
| Gene Symbol: C1qbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007573.2 | NP_031599.2 | 5 - 6 | 763 | 85 | |
| GenBank mRNA | AF300619.1 | - | 5 - 6 | 702 | 85 | |
| AJ001101.1 | - | 5 - 6 | 708 | 85 | ||
| AK010601.1 | - | 5 - 6 | 701 | 85 | ||
| AK010746.1 | - | 5 - 6 | 727 | 85 | ||
| AK011868.1 | - | 5 - 6 | 754 | 85 | ||
| AK169764.1 | - | 5 - 6 | 746 | 85 | ||
| BC038075.1 | - | 5 - 6 | 747 | 3154276 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
immune system process
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
complement activation, classical pathway
RNA splicing
phosphatidylinositol 3-kinase signaling
regulation of complement activation
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
ribosome biogenesis
mature ribosome assembly
positive regulation of apoptotic process
innate immune response
positive regulation of cell adhesion
negative regulation of mRNA splicing, via spliceosome
negative regulation of defense response to virus
positive regulation of protein kinase B signaling
positive regulation of mitochondrial translation
positive regulation of neutrophil chemotaxis
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of trophoblast cell migration
positive regulation of dendritic cell chemotaxis
complement component C1q binding
transcription corepressor activity
mRNA binding
protein kinase C binding
protein binding
hyaluronic acid binding
transcription factor binding
kininogen binding
adrenergic receptor binding
mitochondrial ribosome binding
negative regulation of transcription from RNA polymerase II promoter
immune system process
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
complement activation, classical pathway
RNA splicing
phosphatidylinositol 3-kinase signaling
regulation of complement activation
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
ribosome biogenesis
mature ribosome assembly
positive regulation of apoptotic process
innate immune response
positive regulation of cell adhesion
negative regulation of mRNA splicing, via spliceosome
negative regulation of defense response to virus
positive regulation of protein kinase B signaling
positive regulation of mitochondrial translation
positive regulation of neutrophil chemotaxis
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of trophoblast cell migration
positive regulation of dendritic cell chemotaxis
complement component C1q binding
transcription corepressor activity
mRNA binding
protein kinase C binding
protein binding
hyaluronic acid binding
transcription factor binding
kininogen binding
adrenergic receptor binding
mitochondrial ribosome binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903678 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903677 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903680 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AK169764.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | BC038075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AJ001101.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AK010601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AK010746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AK169764.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | BC038075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AJ001101.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AK010601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P466162 | AK010746.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39345 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121714 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AK169764.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | BC038075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AJ001101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AK010601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AK010746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AK169764.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | BC038075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AJ001101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AK010601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P466162 | AK010746.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27903695 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121714 | - | - |
| Whole Mouse Genome G4122A | A_51_P466162 | AK169764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | BC038075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AJ001101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AK010601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AK010746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AK169764.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | BC038075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AJ001101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AK010601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466162 | AK010746.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137025 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121714 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09418762 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903677 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903678 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27903680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27903593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09676759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09418762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09676771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09414373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27903645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09418760 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388012 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388013 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388016 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388011 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448274_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455821_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388010 | - | - |
| Mouse Genome 430 2.0 Array | 1455821_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448274_at | - | - |
| Mouse Genome 430 2.0 Array | 1445887_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448274_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455821_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1445887_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12261_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12261_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100028854.mm.1 | NM_007573.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028854.mm.1 | NM_007573.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028854.mm.1 | TC1100003118.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448274_at | - | - |
| Mouse Expression Set 430 | 1448274_PM_at | - | - |
| Mouse Expression Set 430 | 1455821_x_at | - | - |
| Mouse Expression Set 430 | 1455821_PM_x_at | - | - |
| Murine Genome U74v2 Set | 96856_at | - | - |
| Murine Genome U74v2 Set | 96856_PM_at | - | - |
| Murine Genome U74v2 Set | 96857_at | - | - |
| Murine Genome U74v2 Set | 96857_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448274_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448274_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455821_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455821_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455821_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448274_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448274_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455821_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 911598 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3170452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225373 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225373 | - | - |
| mouse-6 v1.1 (Illumina) | scl39982.2.14_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39982.2.14_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225373 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225373 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39982.2.14_30-S | - | - |

C1qbp
Genomic Map