| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700022421.mm.1 |
NM_001198949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022421.mm.1 |
NM_001198949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022421.mm.1 |
TC1700002416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022423.mm.1 |
NM_001198949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022423.mm.1 |
NM_001198949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022423.mm.1 |
TC1700002416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022419.mm.1 |
NM_001198949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022419.mm.1 |
NM_001198949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022419.mm.1 |
NM_009067.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022419.mm.1 |
NM_009067.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022419.mm.1 |
TC1700002416.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700022422.mm.1 |
NM_001198949.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022422.mm.1 |
NM_001198949.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700022422.mm.1 |
TC1700002416.mm.1 |
MTA TC |