Assay Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009197.2 | NP_033223.2 | 2 - 3 | 776 | 69 | |
| GenBank mRNA | AB265788.1 | - | 2 - 3 | 602 | 69 | |
| AF045692.1 | - | 2 - 3 | 824 | 69 | ||
| BC080678.1 | - | 2 - 3 | 612 | 6313495 | 69 | |
| BC096587.1 | - | 2 - 3 | 712 | 3992886 | 69 | |
| BM935188.1 | - | 2 - 3 | 289 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a2
Genomic Map