Assay Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009203.3 | NP_033229.3 | 5 - 6 | 1271 | 60 | |
| GenBank mRNA | AB005451.1 | - | 5 - 6 | 1052 | 60 | |
| AK142580.1 | - | 4 - 5 | 420 | 60 | ||
| BC035927.1 | - | 1207 | 4242354 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a12
Genomic Map