| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000014592.mm.1 |
NM_009213.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014592.mm.1 |
NM_009213.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014592.mm.1 |
TC1000002054.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014589.mm.1 |
NM_009213.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014589.mm.1 |
NM_009213.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014589.mm.1 |
TC1000002054.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014590.mm.1 |
NM_009213.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014590.mm.1 |
NM_009213.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014590.mm.1 |
TC1000002054.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014595.mm.1 |
NM_009213.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014595.mm.1 |
NM_009213.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014595.mm.1 |
TC1000002054.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000014594.mm.1 |
NM_009213.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014594.mm.1 |
NM_009213.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000014594.mm.1 |
TC1000002054.mm.1 |
MTA TC |