Assay Details
| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009788.4 | NP_033918.1 | 3 - 4 | 370 | 124 | |
| GenBank mRNA | AK002635.1 | - | 3 - 4 | 372 | 124 | |
| AK005081.1 | - | 3 - 4 | 369 | 124 | ||
| AK005243.1 | - | 3 - 4 | 370 | 124 | ||
| AK028050.1 | - | 3 - 4 | 371 | 124 | ||
| AK028327.1 | - | 3 - 4 | 369 | 124 | ||
| AK038856.1 | - | 371 | 124 | |||
| AK048517.1 | - | 3 - 4 | 371 | 124 | ||
| AK077499.1 | - | 3 - 4 | 369 | 124 | ||
| AK078358.1 | - | 3 - 4 | 368 | 124 | ||
| BC016421.1 | - | 3 - 4 | 301 | 4527000 | 124 | |
| M21531.1 | - | 3 - 4 | 350 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calb1
Genomic Map