Assay Details
| Gene Symbol: Calb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009788.4 | NP_033918.1 | 4 - 5 | 448 | 72 | |
| GenBank mRNA | AK002635.1 | - | 4 - 5 | 450 | 72 | |
| AK005081.1 | - | 4 - 5 | 447 | 72 | ||
| AK005243.1 | - | 4 - 5 | 448 | 72 | ||
| AK028050.1 | - | 4 - 5 | 449 | 72 | ||
| AK028327.1 | - | 4 - 5 | 447 | 72 | ||
| AK038856.1 | - | 449 | 72 | |||
| AK048517.1 | - | 4 - 5 | 449 | 72 | ||
| AK077499.1 | - | 4 - 5 | 447 | 72 | ||
| AK078358.1 | - | 4 - 5 | 446 | 72 | ||
| BC016421.1 | - | 4 - 5 | 379 | 4527000 | 72 | |
| M21531.1 | - | 4 - 5 | 428 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calb1
Genomic Map