Assay Details
| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136059.2 | NP_001129531.1 | 1 - 2 | 86 | 78 | |
| GenBank mRNA | K02588.1 | - | 1 - 2 | 85 | 78 | |
| Y00071.1 | - | 1 - 2 | 86 | 78 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the K02588.1,NM_001136059.2,Y00071.1 transcripts.
Set Membership: |
|

Cyp1a1
Genomic Map