Assay Details
| Gene Symbol: Fjx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010218.2 | NP_034348.2 | 1 | 1159 | 85 | |
| GenBank mRNA | AK051097.1 | - | 1 | 1136 | 85 | |
| BC051990.1 | - | 1 | 1159 | 5687527 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477092 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477089 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477086 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477083 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477088 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477087 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477090 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477094 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477084 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100884 | BC051990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2100884 | NM_010218.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393471 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111663 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100884 | BC051990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2100884 | NM_010218.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21477114 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111663 | - | - |
| Whole Mouse Genome G4122A | A_51_P355589 | BC051990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P355589 | NM_010218.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P355589 | AK051097.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139120 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01970200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02250597 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02250590 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02250592 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477086 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477089 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477087 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477083 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477084 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477088 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477092 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477085 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477090 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21477094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21477103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02090787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01345605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02125906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01611820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01845800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01667072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01737962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02090790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01345606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477112 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02273749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02250591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02250596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01172955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01293673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02002907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01845798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21477134 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10485403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10485404 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450728_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457160_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422733_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10485402 | - | - |
| Mouse Genome 430 2.0 Array | 1457160_at | - | - |
| Mouse Genome 430 2.0 Array | 1450728_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422733_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450728_at | - | - |
| Mouse Genome 430 2.0 Array | 1457160_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422733_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14221_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14221_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200035365.mm.1 | NM_010218.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035365.mm.1 | NM_010218.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035365.mm.1 | TC0200004217.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450728_at | - | - |
| Mouse Expression Set 430 | 1422733_PM_at | - | - |
| Mouse Expression Set 430 | 1422733_at | - | - |
| Mouse Expression Set 430 | 1450728_PM_at | - | - |
| Mouse Expression Set 430 | 1457160_at | - | - |
| Mouse Expression Set 430 | 1457160_PM_at | - | - |
| Murine Genome U74v2 Set | 160816_PM_at | - | - |
| Murine Genome U74v2 Set | 160816_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450728_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450728_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422733_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422733_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422733_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450728_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422733_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450728_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457160_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457160_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 449163 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3390035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3390035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101770722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390035 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737416 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689998 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737416 | - | - |
| mouse-6 v1.1 (Illumina) | scl18949.1_10-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0014221.2_140-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101770722 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0014221.2_140-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101770722 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737416 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689998 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737416 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0014221.2_140-S | - | - |

Fjx1
Genomic Map