Assay Details
| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010761.2 | NP_034891.2 | 9 - 10 | 1013 | 92 | |
| GenBank mRNA | AK154285.1 | - | 9 - 10 | 967 | 92 | |
| AK160205.1 | - | 9 - 10 | 979 | 92 | ||
| AK163150.1 | - | 9 - 10 | 926 | 92 | ||
| AK167345.1 | - | 9 - 10 | 978 | 92 | ||
| BC001984.1 | - | 9 - 10 | 949 | 3489973 | 92 | |
| BC004673.1 | - | 3 - 4 | 433 | 92 | ||
| BC005802.1 | - | 8 - 9 | 1022 | 3707919 | 92 | |
| U50734.1 | - | 9 - 10 | 934 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21578340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21578342 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21578343 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972128 | BC001984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972128 | BC004673.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972128 | BC005802.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972128 | AK154285.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972128 | AK160205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | U50734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | AK160205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | AK163150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | AK167345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | NM_010761.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | U50734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | AK160205.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | AK163150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | AK167345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P154469 | NM_010761.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900010 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628338 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172413 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144243 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | U50734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | AK160205.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | AK163150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | AK167345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | NM_010761.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | U50734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | AK160205.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | AK163150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | AK167345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P154469 | NM_010761.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972128 | BC001984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972128 | BC004673.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972128 | BC005802.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972128 | AK154285.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972128 | AK160205.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21578343 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144243 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04669506 | - | - |
| Whole Mouse Genome G4122A | A_51_P154469 | U50734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | AK160205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | AK163150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | AK167345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | NM_010761.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | U50734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | AK160205.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | AK163150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | AK167345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154469 | NM_010761.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104084 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209382 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209380 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209383 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01691360 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02209371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21578340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21578343 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21578342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21578390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01790113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01847939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01691360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01790118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01249299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01752252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01489095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01790120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01790121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02012785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21578393 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10475279 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475266 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475268 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420745_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10475264 | - | - |
| Mouse Genome 430 2.0 Array | 1420745_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420745_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17151_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17151_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200013910.mm.1 | NM_010761.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200013910.mm.1 | NM_010761.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200013910.mm.1 | TC0200001803.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200013908.mm.1 | NM_010761.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200013908.mm.1 | NM_010761.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200013908.mm.1 | TC0200001803.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200013907.mm.1 | NM_010761.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200013907.mm.1 | NM_010761.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200013907.mm.1 | TC0200001803.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420745_a_at | - | - |
| Mouse Expression Set 430 | 1420745_PM_a_at | - | - |
| Murine Genome U74v2 Set | 101483_at | - | - |
| Murine Genome U74v2 Set | 101483_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420745_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420745_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420745_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420745_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906047 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104920053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040446 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040446 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240178 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234319 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240178 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003372.1_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl20390.11.1_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040446 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104920053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20390.11.1_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040446 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104920053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240178 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240178 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234319 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20390.11.1_18-S | - | - |

Ccndbp1
Genomic Map