Assay Details
| Gene Symbol: Mapkapk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010765.2 | NP_034895.1 | 4 - 5 | 981 | 72 | |
| GenBank mRNA | AF039840.1 | - | 4 - 5 | 995 | 72 | |
| AY533679.1 | - | 4 - 5 | 289 | 72 | ||
| AY533680.2 | - | 4 - 5 | 289 | 72 | ||
| BC019184.1 | - | 4 - 5 | 403 | 5010305 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapkapk5
Genomic Map