Assay Details
| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010769.2 | NP_034899.2 | 3 - 4 | 716 | 83 | |
| GenBank mRNA | AK014342.1 | - | 3 - 4 | 716 | 83 | |
| BC047140.1 | - | 3 - 4 | 683 | 6315509 | 83 | |
| U35035.1 | - | 3 - 4 | 677 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Matn1
Genomic Map