Assay Details
| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010769.2 | NP_034899.2 | 4 - 5 | 840 | 57 | |
| GenBank mRNA | AK014342.1 | - | 4 - 5 | 840 | 57 | |
| BC047140.1 | - | 4 - 5 | 807 | 6315509 | 57 | |
| U35035.1 | - | 4 - 5 | 801 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Matn1
Genomic Map