Assay Details
| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010808.3 | NP_034938.3 | 5 - 6 | 1022 | 97 | |
| GenBank mRNA | AB021226.1 | - | 5 - 6 | 967 | 97 | |
| AJ010262.1 | - | 5 - 6 | 968 | 97 | ||
| CD352405.1 | - | 2 - 3 | 219 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmp24
Genomic Map