Assay Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271407.1 | NP_001258336.1 | 13 - 14 | 1439 | 75 | |
| NM_001271408.1 | NP_001258337.1 | 13 - 14 | 1323 | 75 | ||
| NM_001271409.1 | NP_001258338.1 | 2 - 3 | 304 | 75 | ||
| NM_011366.3 | NP_035496.1 | 13 - 14 | 1326 | 75 | ||
| GenBank mRNA | AB190911.1 | - | 13 - 14 | 1440 | 75 | |
| AF064806.1 | - | 13 - 14 | 1292 | 75 | ||
| AK029703.1 | - | 15 - 16 | 1437 | 75 | ||
| AK164607.1 | - | 1 - 2 | 1768 | 75 | ||
| AK165689.1 | - | 13 - 14 | 1333 | 75 | ||
| BC030933.1 | - | 12 - 13 | 1140 | 4920799 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sorbs3
Genomic Map