Assay Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271407.1 | NP_001258336.1 | 16 - 17 | 1652 | 71 | |
| NM_001271408.1 | NP_001258337.1 | 16 - 17 | 1536 | 71 | ||
| NM_001271409.1 | NP_001258338.1 | 5 - 6 | 517 | 71 | ||
| NM_011366.3 | NP_035496.1 | 16 - 17 | 1539 | 71 | ||
| GenBank mRNA | AB190911.1 | - | 16 - 17 | 1653 | 71 | |
| AF064806.1 | - | 16 - 17 | 1505 | 71 | ||
| AK029703.1 | - | 18 - 19 | 1650 | 71 | ||
| BC030933.1 | - | 15 - 16 | 1353 | 4920799 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sorbs3
Genomic Map