Assay Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271407.1 | NP_001258336.1 | 17 - 18 | 1783 | 64 | |
| NM_001271408.1 | NP_001258337.1 | 17 - 18 | 1667 | 64 | ||
| NM_001271409.1 | NP_001258338.1 | 6 - 7 | 648 | 64 | ||
| NM_011366.3 | NP_035496.1 | 17 - 18 | 1670 | 64 | ||
| GenBank mRNA | AB190911.1 | - | 17 - 18 | 1784 | 64 | |
| AF064806.1 | - | 17 - 18 | 1636 | 64 | ||
| AK029703.1 | - | 19 - 20 | 1781 | 64 | ||
| AK165689.1 | - | 17 - 18 | 1677 | 64 | ||
| BC030933.1 | - | 16 - 17 | 1484 | 4920799 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sorbs3
Genomic Map