Assay Details
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276425.1 | NP_001263354.1 | 1 - 2 | 146 | 55 | |
GenBank mRNA | AJ007310.1 | - | 1 - 2 | 141 | 55 | |
AK007601.1 | - | 1 - 2 | 129 | 55 | ||
AK085730.1 | - | 1 - 2 | 137 | 55 | ||
AK144884.1 | - | 1 - 2 | 137 | 55 | ||
AK153036.1 | - | 1 - 2 | 106 | 55 | ||
AK155956.1 | - | 1 - 2 | 137 | 55 | ||
AK161559.1 | - | 1 - 2 | 131 | 55 | ||
Y15779.1 | - | 1 - 2 | 123 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ007310.1, AK085730.1, NM_001276425.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
