Assay Details
| Gene Symbol: Cops5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277101.1 | NP_001264030.1 | 3 - 4 | 605 | 78 | |
| NM_013715.2 | NP_038743.1 | 3 - 4 | 854 | 78 | ||
| NR_102282.1 | 3 - 4 | 862 | 78 | |||
| GenBank mRNA | AF065386.1 | - | 3 - 4 | 607 | 78 | |
| AF068223.1 | - | 3 - 4 | 622 | 78 | ||
| AK002761.1 | - | 3 - 4 | 636 | 78 | ||
| AK005728.1 | - | 3 - 4 | 641 | 78 | ||
| AK012499.1 | - | 3 - 4 | 641 | 78 | ||
| AK133073.1 | - | 3 - 4 | 634 | 78 | ||
| AK146271.1 | - | 3 - 4 | 641 | 78 | ||
| BC046753.1 | - | 3 - 4 | 600 | 5693100 | 78 | |
| DV056728.1 | - | 3 - 4 | 661 | 78 | ||
| U70736.1 | - | 3 - 4 | 630 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cops5
Genomic Map