Assay Details
| Gene Symbol: Sfrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013834.3 | NP_038862.2 | 2 - 3 | 905 | 80 | |
| GenBank mRNA | AK081052.1 | - | 2 - 3 | 496 | 80 | |
| AK088868.1 | - | 2 - 3 | 907 | 80 | ||
| BC024495.1 | - | 2 - 3 | 875 | 4982195 | 80 | |
| BC094662.1 | - | 2 - 3 | 889 | 30363219 | 80 | |
| U88566.1 | - | 2 - 3 | 878 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sfrp1
Genomic Map