Assay Details
| Gene Symbol: Sult4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013873.3 | NP_038901.3 | 1 - 2 | 323 | 78 | |
| GenBank mRNA | AF059257.1 | - | 1 - 2 | 199 | 78 | |
| AK003034.1 | - | 1 - 2 | 184 | 78 | ||
| AK010293.1 | - | 1 - 2 | 179 | 78 | ||
| AK032195.1 | - | 1 - 2 | 210 | 78 | ||
| AK046368.1 | - | 1 - 2 | 202 | 78 | ||
| AK159034.1 | - | 1 - 2 | 215 | 78 | ||
| BC051132.1 | - | 1 - 2 | 196 | 6531493 | 78 | |
| BC054757.1 | - | 1 - 2 | 323 | 6413084 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sult4a1
Genomic Map