| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200028711.mm.1 |
NM_001164772.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028711.mm.1 |
NM_001164772.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028711.mm.1 |
TC0200003352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028712.mm.1 |
NM_001164772.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028712.mm.1 |
NM_001164772.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028712.mm.1 |
TC0200003352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028713.mm.1 |
NM_001164772.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028713.mm.1 |
NM_001164772.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028713.mm.1 |
TC0200003352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028720.mm.1 |
NM_001164768.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028720.mm.1 |
NM_001164768.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028720.mm.1 |
TC0200003352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164768.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164768.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164770.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164770.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164772.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_001164772.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_013890.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
NM_013890.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028714.mm.1 |
TC0200003352.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164768.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164768.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164769.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164769.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164770.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164770.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164772.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_001164772.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_013890.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
NM_013890.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200028716.mm.1 |
TC0200003352.mm.1 |
MTA TC |