Assay Details
| Gene Symbol: Shoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168505.1 | NP_001161977.1 | 3 - 4 | 1159 | 73 | |
| NM_019658.6 | NP_062632.2 | 3 - 4 | 1200 | 73 | ||
| GenBank mRNA | AF068921.1 | - | 2 - 3 | 835 | 73 | |
| AK146447.1 | - | 3 - 4 | 1197 | 73 | ||
| BC026364.1 | - | 2 - 3 | 4014650 | 73 | ||
| BC049775.1 | - | 2 - 3 | 1065 | 6515905 | 73 | |
| BC083060.1 | - | 3 - 4 | 1159 | 5356514 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Shoc2
Genomic Map