Assay Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019775.3 | NP_062749.2 | 4 - 5 | 444 | 108 | |
| GenBank mRNA | AB021968.1 | - | 4 - 5 | 392 | 108 | |
| AF164524.1 | - | 4 - 5 | 459 | 108 | ||
| AF186188.1 | - | 4 - 5 | 395 | 108 | ||
| AK004045.1 | - | 4 - 5 | 417 | 108 | ||
| BC089577.1 | - | 4 - 5 | 416 | 6766672 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpb2
Genomic Map