Assay Details
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019775.3 | NP_062749.2 | 7 - 8 | 768 | 67 | |
GenBank mRNA | AB021968.1 | - | 7 - 8 | 716 | 67 | |
AF164524.1 | - | 7 - 8 | 783 | 67 | ||
AF186188.1 | - | 7 - 8 | 719 | 67 | ||
AK004045.1 | - | 7 - 8 | 741 | 67 | ||
BC089577.1 | - | 7 - 8 | 740 | 6766672 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
