Assay Details
| Gene Symbol: Cenph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021886.1 | NP_068686.1 | 4 - 5 | 391 | 98 | |
| GenBank mRNA | AB017634.1 | - | 4 - 5 | 391 | 98 | |
| AK006208.1 | - | 4 - 5 | 375 | 98 | ||
| AK011264.1 | - | 4 - 5 | 376 | 98 | ||
| AK011738.1 | - | 4 - 5 | 376 | 98 | ||
| BC025084.1 | - | 4 - 5 | 342 | 3501244 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270035 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382993 | AK042986.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96493 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124026 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116038 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172269 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29270041 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02759368 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116038 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172269 | - | - |
| Whole Mouse Genome G4122A | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | BC025084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | AK011264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | AK011738.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | NM_021886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P492830 | AB017634.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123626 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122913 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116038 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172269 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124026 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10849091 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10870685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10870677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10909536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10909018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10870678 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270034 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270037 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270038 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270036 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270035 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29270039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29270081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11076474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10909018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10867094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11076476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10849091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10699239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10951087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10699240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10649128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29270008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10870679 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10411732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411736 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422016_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10411728 | - | - |
| Mouse Genome 430 2.0 Array | 1422016_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422016_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26886_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26886_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300017325.mm.1 | NM_021886.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017325.mm.1 | TC1300002542.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422016_a_at | - | - |
| Mouse Expression Set 430 | 1422016_PM_a_at | - | - |
| Murine Genome U74v2 Set | 168999_at | - | - |
| Murine Genome U74v2 Set | 168999_PM_at | - | - |
| Murine Genome U74v2 Set | 171383_at | - | - |
| Murine Genome U74v2 Set | 171383_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422016_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422016_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422016_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422016_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906635 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4280097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280097 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693785 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2693785 | - | - |
| mouse-6 v1.1 (Illumina) | scl43589.10.1_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280097 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43589.10.1_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2693785 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2693785 | - | - |

Cenph
Genomic Map