Assay Details
| Gene Symbol: Smoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022315.2 | NP_071710.2 | 8 - 9 | 1034 | 68 | |
| GenBank mRNA | AJ249901.1 | - | 8 - 9 | 1005 | 68 | |
| AK030958.1 | - | 8 - 9 | 1003 | 68 | ||
| AK159749.1 | - | 8 - 9 | 1001 | 68 | ||
| AK160002.1 | - | 8 - 9 | 1001 | 68 | ||
| BC019527.1 | - | 8 - 9 | 981 | 4191849 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smoc2
Genomic Map