Assay Details
| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079235.1 | - | 6 - 7 | 826 | 57 | |
| BC012969.1 | - | 5 - 6 | 715 | 4037243 | 57 | |
| CT010196.1 | - | 4 - 5 | 573 | 57 | ||
| U30509.1 | - | 5 - 6 | 733 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ggt1
Genomic Map