Assay Details
| Gene Symbol: Gnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008140.2 | NP_032166.1 | 3 - 4 | 334 | 69 | |
| GenBank mRNA | AK149254.1 | - | 3 - 4 | 377 | 69 | |
| BC022793.1 | - | 3 - 4 | 334 | 4505478 | 69 | |
| BC051412.1 | - | 3 - 4 | 356 | 6490986 | 69 | |
| BC058810.1 | - | 3 - 4 | 357 | 6490797 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnat1
Genomic Map