Assay Details
| Gene Symbol: Gstt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008185.3 | NP_032211.3 | 2 - 3 | 291 | 82 | |
| GenBank mRNA | AI255817.1 | - | 3 - 4 | 82 | ||
| AK002338.1 | - | 2 - 3 | 291 | 82 | ||
| BC012254.1 | - | 2 - 3 | 206 | 4193816 | 82 | |
| BC055020.1 | - | 2 - 3 | 227 | 4161144 | 82 | |
| BY015234.1 | - | 2 - 3 | 278 | 82 | ||
| W54467.1 | - | 2 - 3 | 242 | 82 | ||
| X98055.1 | - | 2 - 3 | 203 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024841 | AK002338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024841 | BC012254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024841 | BC055020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2024841 | NM_008185.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734966 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868601 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625000 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165843 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160260 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106454 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024841 | AK002338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024841 | BC012254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024841 | BC055020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2024841 | NM_008185.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P179664 | AK002338.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P179664 | BC012254.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P179664 | BC055020.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P179664 | NM_008185.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P179664 | W54467.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258685 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07111286 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106454 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160260 | - | - |
| Whole Mouse Genome G4122A | A_51_P179664 | AK002338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179664 | BC012254.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179664 | BC055020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179664 | NM_008185.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179664 | W54467.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117952 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160260 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08972871 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08258476 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08577429 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08258475 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08258480 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08577428 | - | - |
| Agilent Mouse cDNA Microarray (G4104A) | 367583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08397280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08927039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08258474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08258476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08577429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08397286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08258486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08536714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08536710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08536713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08708384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08217293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08203845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27258713 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370004 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418186_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370000 | - | - |
| Mouse Genome 430 2.0 Array | 1418186_at | - | - |
| Mouse Genome 430 2.0 Array | 1418186_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14871_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14871_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000017057.mm.1 | NM_008185.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017057.mm.1 | NM_008185.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017057.mm.1 | TC1000002383.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017059.mm.1 | NM_008185.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017059.mm.1 | NM_008185.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017059.mm.1 | TC1000002383.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017060.mm.1 | NM_008185.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017060.mm.1 | NM_008185.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017060.mm.1 | TC1000002383.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017058.mm.1 | NM_008185.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017058.mm.1 | NM_008185.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017058.mm.1 | TC1000002383.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 95019_at | - | - |
| Murine Genome U74v2 Set | 95019_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418186_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418186_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 539509 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5290520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290520 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2680531 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2680531 | - | - |
| mouse-6 v1.1 (Illumina) | scl37811.6.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290520 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37811.6.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290520 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2680533 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2680533 | - | - |

Gstt1
Genomic Map