Assay Details
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008489.2 | NP_032515.2 | 2 - 3 | 412 | 56 | |
| GenBank mRNA | AK143562.1 | - | 2 - 3 | 413 | 56 | |
| AK158105.1 | - | 2 - 3 | 295 | 56 | ||
| BC004795.1 | - | 2 - 3 | 300 | 3589072 | 56 | |
| X99347.1 | - | 2 - 3 | 362 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lbp
Genomic Map