Assay Details
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008489.2 | NP_032515.2 | 11 - 12 | 1396 | 61 | |
| GenBank mRNA | BQ176753.1 | - | 4 - 5 | 61 | ||
| X99347.1 | - | 11 - 12 | 1346 | 61 | ||
More Information
Set Membership: |
|

Lbp
Genomic Map