Assay Details
| Gene Symbol: Epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008532.2 | NP_032558.2 | 3 - 4 | 646 | 96 | |
| GenBank mRNA | AK133510.1 | - | 2 - 3 | 328 | 96 | |
| AK145752.1 | - | 2 - 3 | 222 | 96 | ||
| AK167182.1 | - | 2 - 3 | 291 | 96 | ||
| BC005618.1 | - | 2 - 3 | 287 | 3711069 | 96 | |
| BC094465.1 | - | 2 - 3 | 312 | 4948935 | 96 | |
| M76124.1 | - | 2 - 3 | 298 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epcam
Genomic Map