Assay Details
| Gene Symbol: Tcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009337.3 | NP_033363.1 | 3 - 4 | 447 | 89 | |
| NR_110339.1 | 3 - 4 | 695 | 89 | |||
| NR_110340.1 | 2 - 3 | 276 | 89 | |||
| NR_110341.1 | 3 - 4 | 398 | 89 | |||
| GenBank mRNA | AF031956.1 | - | 3 - 4 | 423 | 89 | |
| AK163387.1 | - | 3 - 4 | 430 | 89 | ||
| BC052336.1 | - | 3 - 4 | 428 | 30053730 | 89 | |
| CA558861.1 | - | 3 - 4 | 367 | 89 | ||
| CJ087217.1 | - | 2 - 3 | 258 | 89 | ||
| DV648028.1 | - | 3 - 4 | 654 | 89 | ||
| DV649899.1 | - | 2 - 3 | 247 | 89 | ||
| DV653827.1 | - | 3 - 4 | 447 | 89 | ||
| DV655843.1 | - | 3 - 4 | 391 | 89 | ||
| DV663230.1 | - | 2 - 3 | 149 | 89 | ||
| Y15376.1 | - | 3 - 4 | 423 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960342 | AK163387.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960342 | BC052336.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960342 | AK149361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960342 | NM_009337.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348127 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109585 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960342 | AK163387.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960342 | BC052336.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960342 | AK149361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960342 | NM_009337.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P237129 | AK163387.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P237129 | BC052336.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P237129 | Y15376.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P237129 | AK149361.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P237129 | NM_009337.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28708005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28708002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28708003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28708007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28708010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28708012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707974 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109585 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736612 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02596590 | - | - |
| Whole Mouse Genome G4122A | A_51_P237129 | AK163387.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P237129 | BC052336.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P237129 | Y15376.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P237129 | AK149361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P237129 | NM_009337.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121983 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109585 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09770576 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09770579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10164497 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09770581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09770582 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09770575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10164499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28708026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10089577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09946435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10409785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10151133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09946427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10062715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09770575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09946434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10274484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09765130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28708019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09770581 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10402515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10402518 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422458_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10402514 | - | - |
| Mouse Genome 430 2.0 Array | 1422458_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422458_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21432_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21432_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200016079.mm.1 | NM_001289468.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016079.mm.1 | NM_009337.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016079.mm.1 | NM_009337.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016079.mm.1 | NR_110339.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016079.mm.1 | TC1200002305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016081.mm.1 | NM_001289468.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016081.mm.1 | NM_009337.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016081.mm.1 | NM_009337.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016081.mm.1 | NR_110339.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016081.mm.1 | TC1200002305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016080.mm.1 | NM_001289468.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016080.mm.1 | NM_009337.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016080.mm.1 | NM_009337.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016080.mm.1 | NR_110339.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016080.mm.1 | TC1200002305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200016082.mm.1 | NM_001289468.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016082.mm.1 | NM_009337.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016082.mm.1 | NM_009337.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016082.mm.1 | NR_110339.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200016082.mm.1 | TC1200002305.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422458_at | - | - |
| Mouse Expression Set 430 | 1422458_PM_at | - | - |
| Murine Genome U74v2 Set | 93296_PM_at | - | - |
| Murine Genome U74v2 Set | 93296_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422458_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422458_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422458_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422458_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3520019 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3520019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003520019 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638171 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2638171 | - | - |
| mouse-6 v1.1 (Illumina) | scl42088.4_201-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3520019 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42088.4_201-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2638171 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2985612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2985612 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2638171 | - | - |

Tcl1
Genomic Map