| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100024354.mm.1 |
NM_009581.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024354.mm.1 |
NM_009581.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024354.mm.1 |
TC0100003039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024355.mm.1 |
NM_009581.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024355.mm.1 |
NM_009581.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024355.mm.1 |
TC0100003039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024356.mm.1 |
NM_009581.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024356.mm.1 |
NM_009581.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024356.mm.1 |
TC0100003039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024357.mm.1 |
NM_009581.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024357.mm.1 |
NM_009581.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024357.mm.1 |
TC0100003039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024358.mm.1 |
NM_009581.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024358.mm.1 |
NM_009581.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024358.mm.1 |
TC0100003039.mm.1 |
MTA TC |