Assay Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010369.3 | NP_034499.3 | 6 - 7 | 415 | 92 | |
| GenBank mRNA | AK157851.1 | - | 6 - 7 | 416 | 92 | |
| AK162614.1 | - | 6 - 7 | 400 | 92 | ||
| BC148192.1 | - | 6 - 7 | 679 | 40045496 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | AK162614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | M26385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | NM_010369.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | AK145998.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | AK162614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | M26385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | NM_010369.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77080 | AK145998.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64202 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152624 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130438 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | AK162614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | M26385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | NM_010369.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | AK145998.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | AK162614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | M26385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | NM_010369.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77080 | AK145998.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06569583 | - | - |
| Whole Mouse Genome G4122A | A_52_P77080 | AK162614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | M26385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | NM_010369.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | AK145998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | AK162614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | M26385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | NM_010369.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77080 | AK145998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130995 | AK162614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130995 | BC148192.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130995 | AK157851.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130995 | NM_010369.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130995 | AK145998.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140565 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06569583 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174875 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174881 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174860 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07174874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26000052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06736714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06885707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26000073 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10573060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10573056 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423016_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425643_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10573054 | - | - |
| Mouse Genome 430 2.0 Array | 1423016_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425643_at | - | - |
| Mouse Genome 430 2.0 Array | 1425643_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423016_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14934_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14934_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006633.mm.1 | NM_010369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006633.mm.1 | NM_010369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006633.mm.1 | TC0800000889.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423016_a_at | - | - |
| Mouse Expression Set 430 | 1425643_PM_at | - | - |
| Mouse Expression Set 430 | 1423016_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425643_at | - | - |
| Murine Genome U74v2 Set | 102770_at | - | - |
| Murine Genome U74v2 Set | 102770_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423016_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425643_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425643_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423016_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423016_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425643_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425643_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423016_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 879796 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 540402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000540402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696696 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2696696 | - | - |
| mouse-6 v1.1 (Illumina) | scl33630.8_639-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 540402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33630.8_639-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 540402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2696696 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2696696 | - | - |

Gypa
Genomic Map