| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10390987 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390993 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390992 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390989 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390986 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390991 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390990 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10390994 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1420728_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10390985 |
- |
- |
| Mouse Genome 430 2.0 Array |
1420728_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1420728_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
16670_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
16670_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1100034410.mm.1 |
NM_001159374.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034410.mm.1 |
NM_001159374.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034410.mm.1 |
TC1100003752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034411.mm.1 |
NM_001159374.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034411.mm.1 |
NM_001159374.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034411.mm.1 |
TC1100003752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034412.mm.1 |
NM_001159374.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034412.mm.1 |
NM_001159374.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034412.mm.1 |
TC1100003752.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1420728_at |
- |
- |
| Mouse Expression Set 430 |
1420728_PM_at |
- |
- |
| Murine Genome U74v2 Set |
97767_PM_at |
- |
- |
| Murine Genome U74v2 Set |
97767_at |
- |
- |
| Murine Genome U74v2 Set |
162214_r_at |
- |
- |
| Murine Genome U74v2 Set |
162214_PM_r_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420728_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1420728_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1420728_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1420728_PM_at |
- |
- |