Assay Details
| Gene Symbol: Unc119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011676.2 | NP_035806.1 | 1 - 2 | 262 | 102 | |
| GenBank mRNA | AF030169.1 | - | 1 - 2 | 286 | 102 | |
| AK131955.1 | - | 1 - 2 | 267 | 102 | ||
| BC001990.1 | - | 1 - 2 | 262 | 3484396 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc119
Genomic Map