Assay Details
| Gene Symbol: Siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011734.3 | NP_035864.2 | 1 - 2 | 75 | 81 | |
| GenBank mRNA | AK028598.1 | - | 1 - 2 | 119 | 81 | |
| AK028656.1 | - | 1 - 2 | 75 | 81 | ||
| AK043392.1 | - | 1 - 2 | 96 | 81 | ||
| AK164852.1 | - | 1 - 2 | 82 | 81 | ||
| AK167103.1 | - | 1 - 2 | 91 | 81 | ||
| BC007136.1 | - | 1 - 2 | 93 | 3592286 | 81 | |
| U40408.1 | - | 1 - 2 | 81 | 81 | ||
| U61183.1 | - | 1 - 2 | 74 | 81 | ||
| X98625.1 | - | 1 - 2 | 74 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siae
Genomic Map