Assay Details
| Gene Symbol: Siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011734.3 | NP_035864.2 | 2 - 3 | 240 | 76 | |
| GenBank mRNA | AF156856.1 | - | 3 - 4 | 442 | 76 | |
| AK028598.1 | - | 2 - 3 | 284 | 76 | ||
| AK028656.1 | - | 2 - 3 | 240 | 76 | ||
| AK043392.1 | - | 2 - 3 | 261 | 76 | ||
| AK164852.1 | - | 2 - 3 | 247 | 76 | ||
| AK167103.1 | - | 2 - 3 | 256 | 76 | ||
| BC007136.1 | - | 2 - 3 | 258 | 3592286 | 76 | |
| U40408.1 | - | 2 - 3 | 246 | 76 | ||
| U61183.1 | - | 2 - 3 | 239 | 76 | ||
| X98625.1 | - | 2 - 3 | 239 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF156856.1 transcripts.
Set Membership: |
|

Siae
Genomic Map