Assay Details
| Gene Symbol: Siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011734.3 | NP_035864.2 | 5 - 6 | 816 | 81 | |
| GenBank mRNA | AF156856.1 | - | 6 - 7 | 1018 | 81 | |
| AK028598.1 | - | 5 - 6 | 860 | 81 | ||
| AK028656.1 | - | 5 - 6 | 816 | 81 | ||
| AK033980.1 | - | 64 | 81 | |||
| AK035715.1 | - | 3 - 4 | 550 | 81 | ||
| AK043392.1 | - | 5 - 6 | 837 | 81 | ||
| AK158634.1 | - | 4 - 5 | 754 | 81 | ||
| AK164852.1 | - | 5 - 6 | 823 | 81 | ||
| AK165103.1 | - | 5 - 6 | 843 | 81 | ||
| AK167103.1 | - | 5 - 6 | 832 | 81 | ||
| BC007136.1 | - | 5 - 6 | 834 | 3592286 | 81 | |
| U40408.1 | - | 5 - 6 | 822 | 81 | ||
| U61183.1 | - | 5 - 6 | 815 | 81 | ||
| X98625.1 | - | 5 - 6 | 815 | 81 | ||
More Information
Set Membership: |
|

Siae
Genomic Map