Assay Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289617.1 | NP_001276546.1 | 2 - 3 | 512 | 77 | |
| NM_001289618.1 | NP_001276547.1 | 2 - 3 | 512 | 77 | ||
| NM_001289619.1 | NP_001276548.1 | 2 - 3 | 512 | 77 | ||
| NM_011777.3 | NP_035907.1 | 2 - 3 | 512 | 77 | ||
| GenBank mRNA | AK031415.1 | - | 2 - 3 | 310 | 77 | |
| AK035743.1 | - | 2 - 3 | 283 | 77 | ||
| AK076491.1 | - | 2 - 3 | 283 | 77 | ||
| AK090110.1 | - | 2 - 3 | 279 | 77 | ||
| AK147812.1 | - | 2 - 3 | 513 | 77 | ||
| AK161709.1 | - | 2 - 3 | 308 | 77 | ||
| AK165290.1 | - | 1 - 2 | 446 | 77 | ||
| AK170571.1 | - | 2 - 3 | 286 | 77 | ||
| AK205275.1 | - | 1 - 2 | 91 | 77 | ||
| BC054775.1 | - | 2 - 3 | 471 | 6808149 | 77 | |
| X99063.1 | - | 2 - 3 | 470 | 77 | ||
| Y07711.1 | - | 1 - 2 | 220 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zyx
Genomic Map