Assay Details
| Gene Symbol: Taf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175770.4 | NP_786964.1 | 1 - 2 | 259 | 78 | |
| GenBank mRNA | AF144562.1 | - | 1 - 2 | 206 | 78 | |
| AK029380.1 | - | 1 - 2 | 221 | 78 | ||
| AK075700.1 | - | 1 - 2 | 207 | 78 | ||
| AK076362.1 | - | 1 - 2 | 222 | 78 | ||
| AK089054.1 | - | 1 - 2 | 227 | 78 | ||
| BC029673.1 | - | 1 - 2 | 190 | 5373242 | 78 | |
| DV656353.1 | - | 1 - 2 | 259 | 78 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF144562.1,AK029380.1,AK075700.1,AK076362.1,AK089054.1,BC029673.1,NM_175770.4 transcripts.
Set Membership: |
|

Taf7
Genomic Map